5-(5-Bromo-2-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59401-1G 59401-500MG 59401-5G
76934-798EA 456.19 USD
76934-798 76934-800 76934-802
5-(5-Bromo-2-(trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(5-Bromo-2-(trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂OS
Storage Temperature: Ambient
CAS Number: 2755719-32-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR