5-(2-Bromo-3-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59399-1G 59399-500MG 59399-5G
76934-786EA 593.05 USD
76934-786 76934-788 76934-790
5-(2-Bromo-3-methoxyphenyl)thiazol-2-amine ≥97%
5-(2-Bromo-3-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂OS
MW: 285.16 g/mol
Storage Temperature: Ambient
CAS Number: 2755723-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR