5-(3-Methoxy-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

59395-1G 59395-25G 59395-500MG 59395-5G
76934-772EA 292.81 USD
76934-772 76934-774 76934-776 76934-778
5-(3-Methoxy-4-methylphenyl)thiazol-2-amine ≥97%
5-(3-Methoxy-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 2755718-64-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR