4-Bromo-3-chloro-2-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

59079-1G 59079-25G 59079-500MG 59079-5G
76934-754EA 198.39 USD
76934-754 76934-756 76934-758 76934-760
4-Bromo-3-chloro-2-fluoro-1,1'-biphenyl ≥97%
4-Bromo-3-chloro-2-fluoro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₇BrClF
MW: 285.539 g/mol
Storage Temperature: Ambient
CAS Number: 2755717-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR