5-(2-Fluoro-6-iodophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58895-1G 58895-500MG 58895-5G
76934-740EA 530.45 USD
76934-740 76934-742 76934-744
5-(2-Fluoro-6-iodophenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-6-iodophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆FIN₂S
Storage Temperature: Ambient
CAS Number: 2755718-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR