5-(2-Chloro-5-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58891-1G 58891-500MG 58891-5G
76934-716EA 488.02 USD
76934-716 76934-718 76934-720
5-(2-Chloro-5-methoxyphenyl)thiazol-2-amine ≥97%
5-(2-Chloro-5-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂OS
MW: 240.709 g/mol
Storage Temperature: Ambient
CAS Number: 2755718-20-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR