5-(2-Fluoro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58882-1G 58882-25G 58882-500MG 58882-5G
76934-666EA 396.78 USD
76934-666 76934-668 76934-670 76934-672
5-(2-Fluoro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆F₄N₂S
MW: 262.227 g/mol
Storage Temperature: Ambient
CAS Number: 881402-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR