5-(2-Fluoro-6-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58880-1G 58880-25G 58880-500MG 58880-5G
76934-652EA 396.78 USD
76934-652 76934-654 76934-656 76934-658
5-(2-Fluoro-6-methoxyphenyl)thiazol-2-amine ≥97%
5-(2-Fluoro-6-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂OS
MW: 224.255 g/mol
Storage Temperature: Ambient
CAS Number: 2755722-57-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR