5-(2-(Benzyloxy)-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58687-1G 58687-25G 58687-500MG 58687-5G
76934-626EA 441.34 USD
76934-626 76934-628 76934-630 76934-632
5-(2-(Benzyloxy)-4-methylphenyl)thiazol-2-amine ≥97%
5-(2-(Benzyloxy)-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂OS
MW: 296.387 g/mol
Storage Temperature: Ambient
CAS Number: 2755720-95-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR