5-(2-(Benzyloxy)-5-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58685-1G 58685-500MG 58685-5G
76934-614EA 642.91 USD
76934-614 76934-616 76934-618
5-(2-(Benzyloxy)-5-fluorophenyl)thiazol-2-amine ≥97%
5-(2-(Benzyloxy)-5-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃FN₂OS
MW: 300.351 g/mol
Storage Temperature: Ambient
CAS Number: 2755717-05-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR