5-(4-(Benzyloxy)-3-chlorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

58683-1G 58683-25G 58683-500MG 58683-5G
76934-598EA 429.67 USD
76934-598 76934-600 76934-602 76934-604
5-(4-(Benzyloxy)-3-chlorophenyl)thiazol-2-amine ≥95%
5-(4-(Benzyloxy)-3-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃ClN₂OS
MW: 316.805 g/mol
Storage Temperature: Ambient
CAS Number: 2764728-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR