5-(5-(Benzyloxy)-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58679-1G 58679-500MG 58679-5G
76934-572EA 589.87 USD
76934-572 76934-574 76934-576
5-(5-(Benzyloxy)-2-fluorophenyl)thiazol-2-amine ≥97%
5-(5-(Benzyloxy)-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₆H₁₃FN₂OS
MW: 300.351 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564575
CAS Number: 2734774-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR