5-(3-(Benzyloxy)-4-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58677-1G 58677-500MG 58677-5G
76934-560EA 554.86 USD
76934-560 76934-562 76934-564
5-(3-(Benzyloxy)-4-methylphenyl)thiazol-2-amine ≥97%
5-(3-(Benzyloxy)-4-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂OS
MW: 296.387 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564574
CAS Number: 2734775-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR