5-(4-(Benzyloxy)-2-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58675-1G 58675-500MG 58675-5G
76934-548EA 554.86 USD
76934-548 76934-550 76934-552
5-(4-(Benzyloxy)-2-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-(Benzyloxy)-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₇H₁₆N₂O₂S
MW: 312.386 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564573
CAS Number: 2734775-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR