5-(3-Bromo-5-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58288-1G 58288-500MG 58288-5G
76934-512EA 495.45 USD
76934-512 76934-514 76934-516
5-(3-Bromo-5-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Bromo-5-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂OS
MW: 285.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564550
CAS Number: 2734774-28-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR