5-(2-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58284-1G 58284-500MG 58284-5G
76934-500EA 500.75 USD
76934-500 76934-502 76934-504
5-(2-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Chloro-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆ClF₃N₂S
MW: 278.681 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564547
CAS Number: 2734776-68-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR