5-(4-Fluoro-2-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

58280-1G 58280-500MG 58280-5G
76934-476EA 525.15 USD
76934-476 76934-478 76934-480
5-(4-Fluoro-2-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(4-Fluoro-2-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆F₄N₂S
MW: 262.227 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27874286
CAS Number: 1714903-82-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR