5-(2,3,6-Trifluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57679-1G 57679-500MG 57679-5G
76934-448EA 530.45 USD
76934-448 76934-450 76934-452
5-(2,3,6-Trifluorophenyl)thiazol-2-amine ≥97%
5-(2,3,6-Trifluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂S
Storage Temperature: Ambient
MDL Number: MFCD34564480
CAS Number: 2734776-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR