5-(4-Methoxy-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57440-1G 57440-500MG 57440-5G
76934-410EA 500.75 USD
76934-410 76934-412 76934-414
5-(4-Methoxy-3-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(4-Methoxy-3-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₉F₃N₂OS
MW: 274.262 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530596
CAS Number: 2734775-97-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR