5-(5-Fluoro-2-(methylthio)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57438-1G 57438-500MG 57438-5G
76934-398EA 708.69 USD
76934-398 76934-400 76934-402
5-(5-Fluoro-2-(methylthio)phenyl)thiazol-2-amine ≥97%
5-(5-Fluoro-2-(methylthio)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂S₂
MW: 240.32 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530595
CAS Number: 2734775-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR