5-(6-Bromo-2-fluoro-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57436-1G 57436-500MG 57436-5G
76934-386EA 553.79 USD
76934-386 76934-388 76934-390
5-(6-Bromo-2-fluoro-3-methylphenyl)thiazol-2-amine ≥97%
5-(6-Bromo-2-fluoro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈BrFN₂S
MW: 287.151 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530593
CAS Number: 2734773-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR