5-(2-(Trifluoromethoxy)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

57173-1G 57173-25G 57173-500MG 57173-5G
76934-338EA 233.4 USD
76934-338 76934-340 76934-342 76934-344
5-(2-(Trifluoromethoxy)phenyl)thiazol-2-amine ≥97%
5-(2-(Trifluoromethoxy)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₇F₃N₂OS
MW: 260.236 g/mol
Storage Temperature: Ambient
CAS Number: 926201-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR