4-Iodo-1,5-dimethyl-1H-pyrazole ≥97%

Supplier: AOBChem USA

Warning

57068-1G 57068-25G 57068-5G
76934-324EA 115.64 USD
76934-324 76934-326 76934-328
4-Iodo-1,5-dimethyl-1H-pyrazole ≥97%
4-Iodo-1,5-dimethyl-1H-pyrazole
Formula: C₅H₇IN₂
Storage Temperature: Refrigerator
MDL Number: MFCD15146432
CAS Number: 6647-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR