4-Chloro-2,6-difluorotoluene ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Chloro-1,3-difluoro-2-methylbenzene

56944-1G 56944-25G 56944-500MG 56944-5G
76934-310EA 296 USD
76934-310 76934-312 76934-314 76934-316
4-Chloro-2,6-difluorotoluene ≥95%
4-Chloro-2,6-difluorotoluene
Formula: C₇H₅ClF₂
MW: 162.564 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12922628
CAS Number: 1208076-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR