5-Bromo-2-(methylthio)benzoic acid ≥97%
Supplier: AOBChem USA
|
Warning
|
56899-1G
56899-25G
56899-500MG
56899-5G
76934-294EA
296
USD
76934-294
76934-296
76934-298
76934-300
5-Bromo-2-(methylthio)benzoic acid ≥97%
5-Bromo-2-(methylthio)benzoic acid
Precaución: for Research Use Only
Formula:
C₈H₇BrO₂S MW: 247.109 g/mol Storage Temperature: Ambient |
CAS Number:
62176-40-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...