5-(4-Bromo-2,5-dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56621-1G 56621-500MG 56621-5G
76934-280EA 559.1 USD
76934-280 76934-282 76934-284
5-(4-Bromo-2,5-dimethylphenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2,5-dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrN₂S
MW: 283.187 g/mol
Storage Temperature: Ambient
CAS Number: 2755721-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR