5-(5-Fluoro-2-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56619-1G 56619-500MG 56619-5G
76934-268EA 558.04 USD
76934-268 76934-270 76934-272
5-(5-Fluoro-2-methoxyphenyl)thiazol-2-amine ≥97%
5-(5-Fluoro-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉FN₂OS
MW: 224.255 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12824175
CAS Number: 1182774-94-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR