5-(4-Bromo-2-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56617-1G 56617-500MG 56617-5G
76934-256EA 554.86 USD
76934-256 76934-258 76934-260
5-(4-Bromo-2-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂S
MW: 323.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564395
CAS Number: 2734773-76-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR