5-(4-Bromo-2-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56615-1G 56615-25G 56615-500MG 56615-5G
76934-242EA 385.11 USD
76934-242 76934-244 76934-246 76934-248
5-(4-Bromo-2-methoxyphenyl)thiazol-2-amine ≥97%
5-(4-Bromo-2-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉BrN₂OS
MW: 285.16 g/mol
Storage Temperature: Ambient
CAS Number: 2755724-03-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR