5-(2-Bromo-4-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56608-1G 56608-25G 56608-500MG 56608-5G
76934-200EA 405.27 USD
76934-200 76934-202 76934-204 76934-206
5-(2-Bromo-4-(trifluoromethyl)phenyl)thiazol-2-amine ≥97%
5-(2-Bromo-4-(trifluoromethyl)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₆BrF₃N₂S
MW: 323.132 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564390
CAS Number: 2734773-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR