5-(4-Chloro-2-fluoro-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56221-1G 56221-500MG 56221-5G
76934-174EA 699.14 USD
76934-174 76934-176 76934-178
5-(4-Chloro-2-fluoro-3-methylphenyl)thiazol-2-amine ≥97%
5-(4-Chloro-2-fluoro-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₈ClFN₂S
MW: 242.7 g/mol
Storage Temperature: Ambient
CAS Number: 1872377-49-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR