5-(2,4,6-Trichlorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56154-1G 56154-25G 56154-500MG 56154-5G
76934-160EA 292.81 USD
76934-160 76934-162 76934-164 76934-166
5-(2,4,6-Trichlorophenyl)thiazol-2-amine ≥97%
5-(2,4,6-Trichlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅Cl₃N₂S
Storage Temperature: Ambient
CAS Number: 1593225-23-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR