5-(3-Chloro-5-methoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56138-1G 56138-500MG 56138-5G
76934-148EA 563.34 USD
76934-148 76934-150 76934-152
5-(3-Chloro-5-methoxyphenyl)thiazol-2-amine ≥97%
5-(3-Chloro-5-methoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₀H₉ClN₂OS
MW: 240.709 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564344
CAS Number: 2025861-20-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR