5-(2,4,6-Trifluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56111-1G 56111-25G 56111-500MG 56111-5G
76934-132EA 346.92 USD
76934-132 76934-134 76934-136 76934-138
5-(2,4,6-Trifluorophenyl)thiazol-2-amine ≥97%
5-(2,4,6-Trifluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂S
Storage Temperature: Ambient
MDL Number: MFCD12779911
CAS Number: 1182926-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR