5-(4-Butoxyphenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

56066-1G 56066-25G 56066-500MG 56066-5G
76934-116EA 266.29 USD
76934-116 76934-118 76934-120 76934-122
5-(4-Butoxyphenyl)thiazol-2-amine ≥95%
5-(4-Butoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₆N₂OS
MW: 248.344 g/mol
Storage Temperature: Ambient
CAS Number: 893663-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR