5-(2-Bromo-5-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

56050-1G 56050-500MG 56050-5G
76934-104EA 461.5 USD
76934-104 76934-106 76934-108
5-(2-Bromo-5-fluorophenyl)thiazol-2-amine ≥97%
5-(2-Bromo-5-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrFN₂S
MW: 273.125 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34530480
CAS Number: 1695145-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR