5-(4-Methoxynaphthalen-1-yl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55991-1G 55991-25G 55991-500MG 55991-5G
76934-056EA 363.89 USD
76934-056 76934-058 76934-060 76934-062
5-(4-Methoxynaphthalen-1-yl)thiazol-2-amine ≥97%
5-(4-Methoxynaphthalen-1-yl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂OS
MW: 256.323 g/mol
Storage Temperature: Ambient
CAS Number: 1178322-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR