5-(4-Chloro-2-fluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55955-1G 55955-25G 55955-500MG 55955-5G
76934-024EA 363.89 USD
76934-024 76934-026 76934-028 76934-030
5-(4-Chloro-2-fluorophenyl)thiazol-2-amine ≥97%
5-(4-Chloro-2-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂S
MW: 228.674 g/mol
Storage Temperature: Ambient
CAS Number: 1504848-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR