5-(2,4-Difluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55942-1G 55942-25G 55942-500MG 55942-5G
76934-008EA 292.81 USD
76934-008 76934-010 76934-012 76934-014
5-(2,4-Difluorophenyl)thiazol-2-amine ≥97%
5-(2,4-Difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆F₂N₂S
MW: 212.219 g/mol
Storage Temperature: Ambient
CAS Number: 1178321-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR