5-(4-Iodo-2,6-dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55905-1G 55905-500MG 55905-5G
76933-984EA 628.06 USD
76933-984 76933-986 76933-988
5-(4-Iodo-2,6-dimethylphenyl)thiazol-2-amine ≥97%
5-(4-Iodo-2,6-dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₁IN₂S
MW: 330.188 g/mol
Storage Temperature: Ambient
CAS Number: 1820967-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR