5-(4-Methoxy-3,5-dimethylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

55873-1G 55873-500MG 55873-5G
76933-972EA 530.45 USD
76933-972 76933-974 76933-976
5-(4-Methoxy-3,5-dimethylphenyl)thiazol-2-amine ≥97%
5-(4-Methoxy-3,5-dimethylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.317 g/mol
MDL Number: MFCD24363296
CAS Number: 1556459-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR