5-(3,4,5-Trifluorophenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55865-1G 55865-500MG 55865-5G
76933-960EA 457.25 USD
76933-960 76933-962 76933-964
5-(3,4,5-Trifluorophenyl)thiazol-2-amine ≥97%
5-(3,4,5-Trifluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₅F₃N₂S
MW: 230.21 g/mol
Storage Temperature: Ambient
CAS Number: 1247539-31-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR