4-Bromo-2-fluoro-3-methoxybenzonitrile ≥97%

Supplier: AOBChem USA

Warning

55839-1G 55839-25G 55839-5G
76933-948EA 89.12 USD
76933-948 76933-950 76933-952
4-Bromo-2-fluoro-3-methoxybenzonitrile ≥97%
4-Bromo-2-fluoro-3-methoxybenzonitrile
Formula: C₈H₅BrFNO
MW: 230.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23707401
CAS Number: 1426073-33-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR