5-(4-(Dimethylamino)phenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55739-1G 55739-500MG 55739-5G
76933-906EA 500.75 USD
76933-906 76933-908 76933-910
5-(4-(Dimethylamino)phenyl)thiazol-2-amine ≥97%
5-(4-(Dimethylamino)phenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₃N₃S
MW: 219.306 g/mol
Storage Temperature: Ambient
CAS Number: 1184784-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR