5-(3-Isopropoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55687-1G 55687-25G 55687-500MG 55687-5G
76933-892EA 266.29 USD
76933-892 76933-894 76933-896 76933-898
5-(3-Isopropoxyphenyl)thiazol-2-amine ≥97%
5-(3-Isopropoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₄N₂OS
MW: 234.317 g/mol
Storage Temperature: Ambient
CAS Number: 1509193-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR