5-(2-Ethoxyphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55628-1G 55628-25G 55628-500MG 55628-5G
76933-862EA 296 USD
76933-862 76933-864 76933-866 76933-868
5-(2-Ethoxyphenyl)thiazol-2-amine ≥97%
5-(2-Ethoxyphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 1340170-10-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR