5-(4-Chloro-3-fluorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

55517-1G 55517-25G 55517-500MG 55517-5G
76933-834EA 395.72 USD
76933-834 76933-836 76933-838 76933-840
5-(4-Chloro-3-fluorophenyl)thiazol-2-amine ≥95%
5-(4-Chloro-3-fluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆ClFN₂S
MW: 228.674 g/mol
Storage Temperature: Ambient
CAS Number: 1603213-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR