5-(3-Bromo-4-chlorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

55510-1G 55510-500MG 55510-5G
76933-820EA 465.74 USD
76933-820 76933-822 76933-824
5-(3-Bromo-4-chlorophenyl)thiazol-2-amine ≥95%
5-(3-Bromo-4-chlorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆BrClN₂S
MW: 289.579 g/mol
Storage Temperature: Ambient
MDL Number: MFCD34564303
CAS Number: 1597807-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR