5-(4-Methoxy-3-methylphenyl)thiazol-2-amine ≥97%

Supplier: AOBChem USA

Warning

55503-1G 55503-500MG 55503-5G
76933-806EA 559.1 USD
76933-806 76933-808 76933-810
5-(4-Methoxy-3-methylphenyl)thiazol-2-amine ≥97%
5-(4-Methoxy-3-methylphenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₂N₂OS
MW: 220.291 g/mol
Storage Temperature: Ambient
CAS Number: 1179910-70-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR