5-(3,4-Difluorophenyl)thiazol-2-amine ≥95%

Supplier: AOBChem USA

Warning

55458-1G 55458-500MG 55458-5G
76933-794EA 468.92 USD
76933-794 76933-796 76933-798
5-(3,4-Difluorophenyl)thiazol-2-amine ≥95%
5-(3,4-Difluorophenyl)thiazol-2-amine

Precaución: for Research Use Only

Formula: C₉H₆F₂N₂S
Storage Temperature: Ambient
MDL Number: MFCD12824137
CAS Number: 1182820-77-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR